Learn More
ATCC W6/32 Hybridoma (Mouse, Mus musculus) - ATCC HB-95
Supplier:  ATCC HB95
Encompass
W6/32 is a mouse hybridoma cell line that produces a pan-HLA class I monoclonal antibody reactive with monomorphic determinants on HLA-A, HLA-B, and HLA-C molecules. The antibody is cytotoxic and is commonly used for capture and detection of HLA class I complexes in immunology and immunopeptidomics workflows.
  • Cell line derived from mouse (Mus musculus) hybridoma.
  • Produces a pan-HLA class I monoclonal antibody reactive with HLA-A, HLA-B, and HLA-C.
  • Antibody is IgG2a isotype and exhibits cytotoxic activity.
  • Cross-reacts with ape, monkey, and cow cells; no reactivity with rabbit cells.
  • Used in immunology and immunopeptidomics applications for capture and detection of HLA class I complexes.
Catalog No. 50-238-2397