Learn More
Medchemexpress LLC WAAG-3R TFA | 99.4% | 1644.74 | 10 MG

Supplier: Medchemexpress LLC HYP5419A10MG
WAAG-3R TFA is a biologically active peptide. Aggrecanases, members of the ADAMTS family of proteases, cleave aggrecan, a major structural component of cartilage. Aggrecanase-1 (ADAMTS-4) is a key aggrecanase in human osteoarthritic cartilage. This FRET peptide is utilized in ADAMTS-4 (Aggrecanase-1) and ADAMTS-5 (Aggrecanase-2) assays (Ex/Em = 340/420 nm).
- Powder storage: -80°C for 2 years; -20°C for 1 year
- In solvent storage: -80°C for 6 months; -20°C for 1 month
- Sealed storage, away from moisture and light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.