Learn More
Medchemexpress LLC WAAG-3R TFA | 99.8% | 1644.74 | 5 MG

Supplier: Medchemexpress LLC HYP5419A5MG
WAAG-3R TFA is a biologically active peptide. Aggrecanases, a family of ADAMTS proteases, cleave aggrecan, a major structural component of cartilage. Aggrecanase-1 (ADAMTS-4) is notably present in human osteoarthritic cartilage. This FRET peptide is suitable for use in ADAMTS-4 (Aggrecanase-1) and ADAMTS-5 (Aggrecanase-2) assays.
- Biologically active peptide
- Used in ADAMTS-4 and ADAMTS-5 assays
- FRET peptide with Ex/Em = 340/420 nm
- Soluble in H2O at 12.5 mg/mL (ultrasonic required)
- Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture and light)
- In-solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.