Learn More
Electron Microscopy Sciences Single and Multi Wafer Containers, Use: 6" (150 mm). Material: Polypropylene. Dimensions: 160 x 11 mm., 6/PK
SDP
Supplier:  Electron Microscopy Sciences 649176
Encompass_Preferred
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring. Boxes are transparent and made from Anti-Electrostatic Polycarbonate. Use: 6" (150 mm). Material: Polypropylene . Dimensions: 160 x 11 mm .
Catalog No. 50-209-0887