Learn More
Electron Microscopy Sciences Rubis Wafer Tweezers Style 43LB-4 Peek, 138 mm
SDP
Supplier:  Electron Microscopy Sciences 7824001
Encompass_Preferred
For wafers handling. Made of PEEK an ESD safe soft, conductive fiber, with heat resistance up to 250°C. Clean-room compatible.
OAL: 138 mm
Width: 13 mm
Catalog No. 50-239-38