Learn More
New England Biolabs, Inc. WarmStart Afu Uracil DNA Glycosylase (UDG) - 200 units
SDP
Supplier:  New England Biolabs, Inc. M1282S
Encompass
A thermostable homolog of the E. coli Uracil-DNA Glycosylase (UDG) (1,2) from Archaeoglobus fulgidus. WarmStart Afu UDG catalyzes the release of free uracil from uracil-containing DNA. It is formulated with a reversibly-bound aptamer, which inhibits its activity at temperatures below 37°C. WarmStart Afu UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA.
Catalog No. 50-258-1808