Learn More
Seracare Life Sciences Inc WASH SOLUTION CONCENTRATE
Supplier:  Seracare Life Sciences Inc 51500011
Encompass_Preferred
Convenient 20X concentrate containing imidazole-buffered saline and Tween 20. Product contains 1 x 1L bottle, enough to prepare 20 liters of working solution. Suitable for ELISA, blotting and immunohistochemistry applications.
Catalog No. 50-674-11