Learn More
M2 Polymer Technologies Inc WASTE LOCK 770 25LB
SDP
Supplier:  M2 Polymer Technologies Inc WASTELOCK77025LB
Encompass_Preferred
A 25 lbs pail of a cross-linked Sodium Polyacryalte Superabsorbent polymer. Product absorbs as much as 500 times its weight in DI water. Particle size 200 - 800 um.
Catalog No. NC0287027