Learn More
Medchemexpress LLC Water-17O | 13768-40-6 | 99.0% | 19.02 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY108872S110MG
Encompass_Preferred
Water-17O is the 17O labeled water, primarily intended for research use only. This compound is not sold to patients. It is categorized under isotope-labeled compounds, which are stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules. These compounds often serve as tracers for quantitation during drug development processes. Deuteration, for instance, has shown potential in influencing the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-45776