Learn More
Medchemexpress LLC Water-17O | 13768-40-6 | 99.0% | 19.02 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY108872S1100MG
Encompass_Preferred
Water-17O is the 17O labeled water. It can be used as a tracer and as an internal standard for quantitative analysis. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes, like 17O, are incorporated into drug molecules as tracers for quantitation during drug development
Catalog No. 50-003-46106