Learn More
Medchemexpress LLC Water-17O | 13768-40-6 | 99.0% | 19.02 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY108872S1
Encompass_Preferred
Water-17O is the 17O labeled water. It is intended for research use only.
  • Labeled with 17O isotope.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
Catalog No. 50-003-47225