Learn More
Medchemexpress LLC Water-18O | 14314-42-2 | 99.99% | 20.02 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY108872S100MG
Encompass_Preferred
Water-18O is the 18O-labeled Water. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development. Deuteration has also shown potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-44693