Learn More
Sigma Aldrich Fine Chemicals Biosciences Water-18O | 14314-42-2 | MFCD00064503 | 1g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3298781G
Encompass
Water-18O | Putity: 97 atom % 18O | MW: 20.02 | 14314-42-2 | MFCD00064503 | 1g
Catalog No. NC1603437