Learn More
Yamato Scientific America, Inc. Water purification system in 220V standard equipped with membrane filter and a 100L tank. This system produces Type 1 deionized water and Type 2 distilled water.
Supplier:  Yamato Scientific America, Inc. WG1012
Encompass_Preferred
Yamato’s Auto Still WG-1012 multi-purpose water purification system comes standard with water intake membrane filters to increase the life span of all consumables and a 100L storage tank capacity. The standard equipped membrane filters at feeding port protect pure water production from contamination. WG-1012 produces ASTM deionized water type 1 and type 2 distilled water.
Catalog No. NC2334157
Add to Cart
Request a Quote