Learn More
Yamato Scientific America, Inc. Benchtop water purification system in 115V standard equipped with membrane filter and a 30L tank . This system produces Type 1 deionized water and Type 2 distilled water
Supplier:  Yamato Scientific America, Inc. WG252115V
Encompass_Preferred
Yamato’s Auto Still WG-252 multi-purpose water purification system comes standard with water intake membrane filters to increase the life span of all consumables. The standard equipped membrane filters at feeding port protect pure water production from contamination. It produces ASTM deionized water Type 1 and Type 2 distilled water kept in a 30L storage tank.
Catalog No. NC2334078