Learn More
Microtech Scientific WATER SAMPLE BOTTLES W/ PILLS
Supplier:  Microtech Scientific MS5R120PS
Encompass_Preferred
Water Sample Bottles, 120mL Polystyrene, with Sodium Thiosulfate and STB (Breakaway) Tamper Caps, Sterile, 100/Box, 200/Case
Catalog No. NC0369379