Learn More
Hanna Pharmaceutical Supply Co., Inc. STERILE WATER INJ 20ML
Supplier:  Hanna Pharmaceutical Supply Co., Inc. 0409488720
Encompass_Preferred
20ml Fliptop Vial Sterile, nonpyrogenic, solute-free preparation of distilled water for injection. For use only as a sterile solvent or diluent vehicle for drugs or solutions suitable for parenteral administration pH 5.5 (5.0-7.0). Contains no bacteriostat, antimicrobial agent or added buffer 25/PK
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
Catalog No. NC9013644