Learn More
Waters Corp Waters PAH C18 Column, 120 Å, 5 μm, 2.1 mm, 150 mm
Supplier:  Waters Corp 186001261
Encompass_Preferred
This PAH C18 Column is optimized for HPLC analysis of Polyaromatic Hydrocarbons (PAHs), frequently monitored environmental contaminants. It meets standard methods for PAH analysis in various matrices including air, water, waste, and food. A simple water/acetonitrile gradient achieves baseline resolution and excellent peak symmetry for target PAH analytes.
  • Optimized for PAH analysis in HPLC.
  • High resolving power with baseline resolution and peak symmetry for target PAH analytes.
  • Robust manufacturing in a cGMP, ISO 9001-certified facility.
  • Quality assurance via chromatographic testing of each batch for repeatable performance.
Catalog No. 50-780-633