Learn More
Waters Corp Waters Column, C18, 5 μm, 120 Å, 3 mm x 250 mm
Supplier:  Waters Corp 186001263
Encompass_Preferred
Polyaromatic hydrocarbons (PAHs) are frequently monitored environmental contaminants. Standard and official methods for PAH analysis often specify HPLC, usually with UV and fluorescence detection, as the recommended analytical procedure. This column is designed for the HPLC analysis of PAHs.
  • Allows for baseline resolution and exceptional peak symmetry for all target PAH analytes in a short time, using a simple water/acetonitrile mobile phase.
  • Provides superior peak identification and quantitation for PAHs.
  • Produced with ultra-pure reagents in a cGMP, ISO 9001-certified facility.
  • Each batch of packing material is chromatographically tested with acidic, basic, and neutral analytes.
  • Results are held to limited specification ranges to ensure good, repeatable performance.
Catalog No. 50-780-635