Learn More
Waters Corp Waters Positive Pressure Processor, 96-Well, SPE, 80 psi
Supplier:  Waters Corp 186006961
Encompass_Preferred
This Positive Pressure-96 Processor is engineered for 96-well and 1 cc flangeless cartridge formats, providing advanced operation for solid-phase extraction and filtration. It maintains constant pressure even with partially filled plates, enhancing reproducibility and ease of use. The processor automatically adjusts to various plate heights, accommodating all 96-well plate formats.
  • Utilizes positive pressure for efficient sample preparation and reproducible results.
  • Maintains constant pressure across 96 wells, even partially filled.
  • Fine control over flow via adjustable inert gas.
  • Accommodates pressures up to 80 psi, ideal for viscous samples.
  • Reduces well-plugging risk with rapid gas flow.
  • Designed for 96-well and 1 cc flangeless cartridge formats.
  • Self-adjusting for compatibility with all 96-well plate heights.
Catalog No. 50-596-914
Only null left
Add to Cart
Request a Quote