Learn More
Waters Corp Waters Spherisorb C8 Column, 80Å, 10 μm, 4.6 mm × 250 mm
Supplier:  Waters Corp PSS832815
Encompass_Preferred
Spherisorb C8 columns are general-purpose, silica-based reversed-phase columns that contain an Octyl ligand. These USP L7 columns are used when a less retentive sorbent is required compared to Spherisorb ODS columns.
  • General purpose, silica-based reversed-phase column
  • Less retentive sorbent compared to Spherisorb ODS columns
  • Widely referenced in scientific literature with a wide range of validated methods
  • High-quality bonded phases for various separation selectivities
  • Industry-standard Parker-style column end fittings
  • Durable and highly efficient, offering flexibility in method optimization and solvent consumption reduction
  • Manufactured under cGMP guidelines in an ISO 9002 certified and FDA registered plant
  • Individually tested with a test chromatogram supplied
Catalog No. 50-784-454