Learn More
Waters Corp Waters Corporation Column, C8, 5 μm, 80 Å, 4 mm x 250 mm
Supplier:  Waters Corp PSS845281
Encompass_Preferred
Spherisorb C8 columns are general-purpose, silica-based, reversed-phase columns featuring an Octyl ligand. As USP L7 columns, they offer high efficiency and durability, ideal for applications requiring a less retentive sorbent than ODS columns.
  • Manufactured in controlled chemistry plants to satisfy cGMP and ISO 9002 standards.
  • Ensures critical performance features such as peak symmetry, extended column lifetime, high efficiency, and excellent column-to-column reproducibility.
  • Designed to optimize column stability, efficiency, and symmetry by addressing critical process parameters like packaging pressure, temperature, flow rate, slurry volume, and solvent choice.
  • Supports a particle size of 5 μm and sustains a maximum pressure of 6000 psi (415 Bar).
Catalog No. 50-784-665