Learn More
Waters Corp Spherisorb Cyano (CN) Column, Normal Phase, 80Å, 5 µm, 4.6 mm X 150 mm, 1/pk
Supplier:  Waters Corp PSS830913
Encompass_Preferred
The Spherisorb CN chemistry is general purpose normal phase propyl cyano bonded ligand to a silica substrate. This column is conditioned and shipped with normal phase solvent.
Catalog No. 50-784-341