Learn More
Waters Corp Spherisorb Amino (NH2) Column, 80Å, 5 µm, 4.6 mm X 250 mm, 1/pk
Supplier:  Waters Corp PSS831115
Encompass_Preferred
Spherisorb NH2 (Amino) columns contain a silica-based aminopropyl bonded sorbent. The column is listed as USP L8 and is used for anion-exchange, normal-phase and HILIC separations.
Catalog No. 50-784-363