Learn More
Waters Corp Waters Column, Amino (NH2), 10 μm, 4.6 mm ID, 250 mm
Supplier:  Waters Corp PSS833615
Encompass_Preferred
These columns contain a silica-based aminopropyl bonded sorbent. Classified as USP L8, they are suitable for anion-exchange, normal-phase, and HILIC separations. Manufactured in accordance with cGMP and ISO 9002 guidelines, they utilize pure chemicals and undergo various tests.
  • Achieve optimal peak symmetry, high efficiency, and excellent column stability.
  • Analyze impacts of process variables including slurry solvent selection, packing pressure duration, temperature, flow rate, and slurry volume.
  • Consistently achieve quality requirements for liquid chromatography.
  • Offer exceptional flexibility in equipment optimization and lower solvent use.
  • Provide important performance characteristics such as column efficiency, peak symmetry, column longevity, and column-to-column consistency.
Catalog No. 50-784-485