Learn More
Waters Corp Waters Corporation Chromatography Column, C18, 80Å, 3 μm, 2 mm x 100 mm
Supplier:  Waters Corp PSS833422
Encompass_Preferred
These general purpose, silica-based, reversed-phase C18 columns feature low ligand density with a specified carbon load of 6.2%.
  • General purpose, silica-based, reversed-phase C18 column
  • Low ligand density
  • Specified carbon load of 6.2%
Catalog No. 50-784-475