Learn More
Waters Corp Spherisorb SAX Column, 80Å, 5 µm, 4 mm X 125 mm, 1/pk
Supplier:  Waters Corp PSS845304
Encompass_Preferred
Spherisorb SAX columns contain a silica-based quaternary ammonium bonded sorbent. The column is listed as USP L14 and is used for anion-exchange separations.
Catalog No. 50-784-687