Learn More
Waters Corp Waters Silica Column, 80Å, 5 μm, 4 mm x 250 mm
Supplier:  Waters Corp PSS845540
Encompass_Preferred
The Spherisorb silica chemistry provides a general-purpose unbonded silica substrate for normal phase applications. These packing materials are produced in a cGMP, ISO 9001-certified facility using ultra-pure reagents for reliable lab equipment.
  • Better efficiency, peak shape, reproducibility, and lifetime.
  • Created expressly for use with UPLC systems.
  • Each batch is chromatographically tested with acidic, basic, and neutral analytes for repeatable performance.
  • Performance chromatogram and certificate of batch analysis provided on intelligent chip.
  • Wide variety of particle sizes and chemistries available for method development.
Catalog No. 50-784-696