Learn More
Waters Corp Spherisorb Silica Column, 80Å, 5 µm, 1 mm X 150 mm, 1/pk
Supplier:  Waters Corp PSS830133
Encompass_Preferred
The Spherisorb silica chemistry is general purpose unbonded silica substrate designed for normal phase applications.
Catalog No. 50-784-299