Learn More
Waters Corp XBridge Column, 125Å SEC, MaxPeak Premier, 2.5 μm
Supplier:  Waters Corp 186011355
Encompass_Preferred
This column is designed for scientists working with biotherapeutic peptides, such as GLP receptor agonists and insulin-like peptides, and small proteins ranging from approximately 1,000 to 80,000 Dalton. It provides higher resolution of size variants, more reproducible high molecular weight protein (HMWP) quantitation, reduced run times, and solvent cost savings. Utilizing MaxPeak High Performance Surface (HPS) Technologies and innovative manufacturing, it minimizes non-desired secondary ionic interactions between biomolecules and column hardware surfaces.
  • Higher resolution of size variants.
  • More reproducible HMWPs quantitation.
  • Reduced run times.
  • Solvent cost savings.
  • Minimizes non-desired secondary ionic interactions.
  • Features a hybrid particle substrate.
  • Offers a wide pH range from 2.5 to 8 pH.
  • Supports a molecular weight range of 1,000 to 80,000.
Catalog No. NC3706553