Learn More
ECM Bioscience WAVE Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience WK6130
Encompass_Preferred
The WAVE phospho-regulation antibody sampler kit can be used to examine phosphorylation of WAVE1 at Tyr-125 and Tyr-150, as well as the conserved sites in WAVE2 and WAVE3. The kit also includes polyclonal antibodies to monitor total expression levels of WAVE1 and WAVE2. Applications: WB, E; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8875