Learn More
Medchemexpress LLC Way-169916 | 669764-18-5 | 99.3% | 334.29 g·mol⁻¹ | C17H13F3N2O2 | 50 MG

Supplier: Medchemexpress LLC HY11772650MG
WAY-169916 is a pathway-selective ligand of the estrogen receptor (ER) that inhibits NF-κB transcriptional activity and exhibits anti-inflammatory effects. It is supplied as a solid research compound for laboratory use and has defined solubility and storage recommendations to preserve stability.
- High purity (99.3%) suitable for research applications.
- Solid powder format for ease of handling and long-term storage.
- Soluble in DMSO at 125 mg/mL; ultrasonic assistance and freshly opened DMSO recommended.
- Stable when stored as powder at -20°C for up to 3 years.
- Recommended storage in solvent: -80°C for up to 6 months.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.