Learn More
Medchemexpress LLC HY-101271 5mg Medchemexpress, WAY-200070 CAS:440122-66-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012715MG
Encompass_Preferred
Medchemexpress, HY-101271 5mg WAY-200070 CAS:440122-66-7 WAY-200070 is a selective estrogen receptor β ( ERRβ ) agonist with an IC 50 of 2.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2081