Learn More
Apexbio Technology LLC WAY-262611 1123231-07-1 10mg
SDP
Supplier:  Apexbio Technology LLC B481810
Encompass_Preferred
WAY-262611 (CAS 1123231-07-1) is a small molecule agonist of the Wnt/ -catenin signaling pathway It enhances bone formation by activating -catenin-dependent signaling and antagonizing the activity of Dickkopf-1 (Dkk1) an endogenous Wnt pathway inhibitor In TCF-luciferase reporter assays WAY-262611 demonstrates an EC50 of 0 63 M indicating potent activation of Wnt signaling Due to its capacity to modulate osteogenic pathways WAY-262611 is widely utilized in studies exploring bone metabolism and related disorders
Catalog No. 50-224-0377