Learn More
Medchemexpress LLC WAY-600 | 1062159-35-6 | C28H30N8O | 2 MG
SDP
Supplier:  Medchemexpress LLC HY152722MG
Encompass_Preferred
WAY-600 is a potent, ATP-competitive, and selective mTOR inhibitor. It blocks mTOR complex 1/2 (mTORC1/2) assembly and activation, exhibiting concentration and time-dependent inhibition of HepG2 and Huh-7 cell viability. It also inhibits BrdU incorporation and increases caspase-3 and caspase-9 activity in HepG2 cells, and inhibits HepG2 tumor growth in nude mice.
  • Potent, ATP-competitive, and selective mTOR inhibitor.
  • Blocks mTOR complex 1/2 (mTORC1/2) assembly and activation.
  • Inhibits HepG2 and Huh-7 cell viability.
  • Decreases HepG2 cell colonies and inhibits BrdU incorporation.
  • Increases caspase-3 and caspase-9 activity.
  • Inhibits HepG2 tumor growth in nude mice.
Catalog No. 50-003-72101