Learn More
Grainger Lubricant, -60°F to 300 Degrees F, No Additives, 11 oz. Aerosol Can
Supplier:  Grainger 20JY61
Encompass_Preferred
WD-40 helps to penetrate, loosen, and free stuck, frozen, or rusted parts. It also helps to protect metal against corrosive elements as it cleans and removes grease, grime, tar, and more. WD-40 displaces moisture and is nonconductive. Min. Operating Temp.-60 Degrees F Max. Operating Temp.300 Degrees F.
Catalog No. NC0933267