Learn More
Medchemexpress LLC WDR91-IN-1 | 2759614-35-0 | 98.9% | 372.85 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY161017
Encompass_Preferred
WDR91-IN-1 (Compd 1) is a first-in-class small-molecule ligand for WDR91, with a KD of 6 μM.
  • First-in-class small-molecule ligand for WDR91
  • KD of 6 μM
Catalog No. 50-003-65673