Learn More
MSC Lift-All Endless Web Sling, Polyester, 12 ft., 4 in. Width, 11,500 lb. Capacity, 1 ea.
Supplier:  MSC 87843272
Encompass
This Type 5 endless web sling is designed for material lifting applications, providing reliable load control with its durable polyester construction. Featuring a 1-ply design, this sling offers a 11,500 lb. vertical capacity, 9,200 lb. choker capacity, and 23,000 lb. basket capacity. It is built for moderate duty and includes features to aid in inspection and improve service life.
  • Polyester material provides less stretch and better load control
  • Red core yarns visually reveal damage for easier inspection
  • Abrasion-resistant polymer tag included for traceability
  • Yellow treatment enhances abrasion resistance and sling life
  • Meets or exceeds OSHA 1910.184, ASME B30.9 and WSTDA standards
  • Maximum operating temperature of 200°F
Catalog No. 50-005-47749