Learn More
Medchemexpress LLC Wedelolactone | 524-12-9 | 314.25 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN055110MM/1ML
Encompass_Preferred
Wedelolactone suppresses LPS-induced caspase-11 expression by directly inhibiting the IKK Complex. It also inhibits 5-lipoxygenase (5-Lox) with an IC50 of 2.5 μM. Wedelolactone induces caspase-dependent apoptosis in prostate cancer cells via downregulation of PKCε without inhibiting Akt. It can be extracted from Eclipta alba and is used for cancer research.
  • Suppresses LPS-induced caspase-11 expression
  • Inhibits 5-lipoxygenase (5-Lox)
  • Induces caspase-dependent apoptosis in prostate cancer cells
  • Downregulates PKCε without inhibiting Akt
  • Suitable for cancer research
Catalog No. 50-003-88423