Learn More
Medchemexpress LLC HY-N0551 5mg Medchemexpress, Wedelolactone CAS:524-12-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN05515MG
Encompass_Preferred
Medchemexpress, HY-N0551 5mg Wedelolactone CAS:524-12-9 Wedelolactone, a natural product from Ecliptae herba, suppresses LPS-induced caspase-11 expression by directly inhibiting the IKK Complex. Wedelolactone inhibits 5-lipoxygenase (5-Lox) (IC50~2.5 μM) activity by an oxygen radical scavenging mechanism. Wedelolactone induces caspase-dependent apoptosis in prostate cancer cells via downregulation of PKCε without inhibiting Akt. Anti-cancer, anti-inflammatory, and antioxidant activities[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0284