Learn More
Selleck Chemical LLC Wedelolactone S9042-5MG
SDP
Supplier:  Selleck Chemical LLC S90425MG
Encompass_Preferred
Wedelolactone a medicinal plant-derived natural compound is an inhibitor of IKK that is critical for activation of NF- B by mediating phosphorylation and degradation of I B Wedelolactone is also an inhibitor of caspase-11
Catalog No. 50-000-23699