Learn More
Medchemexpress LLC HY-138239 5mg Medchemexpress, WEE1-IN-3 CAS:2272976-28-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1382395MG
Encompass_Preferred
Medchemexpress, HY-138239 5mg WEE1-IN-3 CAS:2272976-28-8 WEE1-IN-3 is a potent Wee1 kinase inhibitor with an IC50 of <10 nM. WEE1-IN-3 has anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1519