Learn More
ABclonal Technology WEE1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A22275100UL
Encompass_Preferred
This gene encodes a nuclear protein which is a tyrosine kinase belonging to the Ser Thr family of protein kinases This protein catalyzes the inhibitory tyrosine phosphorylation of CDC2 cyclin B kinase and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase
Catalog No. 50-000-07060