Learn More
Apexbio Technology LLC WEHI-539 hydrochloride, 5mg. Cas: 2070018-33-4 MFCD: MFCD26960962. Bcl-xL inhibitor,high affinity and selective
SDP
Supplier:  Apexbio Technology LLC A86345
Encompass_Preferred
WEHI-539 hydrochloride is a small-molecule inhibitor of BCL XL with IC50 value of 1.1 nM [1].WEHI-539 was designed as a BCL-XL inhibitor with high affinity. It interacted with the binding groove of BCL-XL with Kd value of 0.6 nM Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5170