Learn More
Medchemexpress LLC WEHI-9625 | 2595314-46-6 | 99.5% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY128777
Encompass_Preferred
WEHI-9625 is a tricyclic sulfone, first-in-class inhibitor of apoptosis with an EC50 of 69 nM. It binds to VDAC2, promoting its ability to inhibit apoptosis driven by mouse BAK. It is completely inactive against both human BAK and the related apoptosis effector BAX. Intended for research use only.
  • Tricyclic sulfone structure
  • First-in-class inhibitor of apoptosis
  • Binds to VDAC2
  • Promotes VDAC2's ability to inhibit apoptosis driven by mouse BAK
  • Inactive against human BAK and BAX
Catalog No. 50-003-42400