Learn More
Electron Microscopy Sciences Weigert's Iron Hematoxylin B 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2635003
Encompass_Preferred
Used in Van Gieson's Method for Collagen Fibers for collagen, muscle and cornified epithelium. See references Clark, G., (ed) Staining Procedures, 3rd Ed., Williams & Wilkins Co., Baltimore, c. 1973, p. 50, Luna, L (ed) AFIP Manual o f Histological Staining Methods, 3rd ED., New York: McGraw Hill Publications, c 1968, p.76, Van Gieson, I., New York Med . J., 50:57-60, 1889, and Mallory, F.B., Pathological Technique, Hafner Publishing., New York, c. 1961, p. 152.
Catalog No. 50-317-59