Learn More
Supply Solutions Weigerts iron hematoxylin solution, MFCD00284879, 1 set
Supplier:  Supply Solutions SIGMA HT10791SET
Encompass_Preferred
Weigert's iron hematoxylin solution set consists of HT107 Part A and HT109 Part B. Recommended in lengthy or acidic stain procedures. Resists decolorization better than aluminum based hematoxylin formulations.
Catalog No. NC0829134