Learn More
Thomas Scientific Pour-Boat Weighing Dish, Anti-Static, White, Medium (83 x 132 x 26mm), 500/pk
SDP
Supplier:  Thomas Scientific 1163Z51
Encompass
labForce flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss. Pour-spout design easily dispenses samples without flexing the dish. Flat-bottom dishes are made from 12-mil thick polystyrene for better strength, stability, and handling.
Catalog No. NC2006763