Learn More
MSC Weiler Abrasives Flue Brush, 2-1/2 in Diameter, Steel Fill, Double Spiral
Supplier:  MSC 82071945
Encompass
This 2-1/2 inch double spiral flue brush is designed for internal cleaning of pipes and flues. It features a durable steel fill with a 0.012 inch diameter and is fitted with a 1/4 x 2 inch NPT pipe nipple.
  • Fitted with 1/4 x 2 in NPT pipe nipple.
  • Ideal for internal cleaning of pipes and flues.
  • Double stem double spiral design.
Catalog No. 50-283-8605