Learn More
WELCH MATERIALS INC Welchrom NH2 SPE Cartridges, 200mg/3mL, 50pcs/pk
Supplier:  WELCH MATERIALS INC 0050911004
Encompass_Preferred
SPE Cartridges, Welchrom NH2, 200mg/3mL, 50pcs/pk, brand: Welchrom, bonding phase: Aminopropyl, particle size: 40-60 μm, surface area: 480 m2/g, pore size: 60Å, Pore volume: 0.80 cm3/g, carbon loading: 3.5%, endcapped.
Catalog No. NC2053866